Aminopyrazine, 99+%, Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
CAS: 5049-61-6 | C4H5N3 | 95.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number ACR104490500
Price (MYR)
2,246.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,246.00
Add to cart
Chemical Identifiers
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Alkalinity=<0.3 ml/g (ml 0.1 N HCl/g)
Arsenic (As)=<5 ppm
Appearance (Color)White to faintly yellowish-white
Cadmium (Cd)=<10 ppm