Aminopyrazine, 99+%, Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
CAS: 5049-61-6 | C4H5N3 | 95.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number ACR104490100
Price (MYR)
679.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
679.00
Add to cart
Chemical Identifiers
CAS3301-79-9
IUPAC Name3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-6-carboxylic acid
Molecular FormulaC21H12O7
InChI KeyBZTDTCNHAFUJOG-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2C(=O)OC3(C2=C1)C1=CC=C(O)C=C1OC1=CC(O)=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
HPLC>=95 %
Appearance (Color)Yellow to orange
Appearance (Form)Powder