3-Amino-1-propanol, 99%, Thermo Scientific Chemicals
3-Amino-1-propanol, 99%, Thermo Scientific Chemicals
3-Amino-1-propanol, 99%, Thermo Scientific Chemicals
3-Amino-1-propanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Amino-1-propanol, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR104465000500 mLGlass bottle
Catalog number ACR104465000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2943-75-1
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4135-1.4195 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥94.0%