4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Catalog number ACR104365000
Price (MYR)
1,336.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,336.00
Add to cart
Chemical Identifiers
CAS78-69-3
IUPAC Name3,7-dimethyloctan-3-ol
Molecular FormulaC10H22O
InChI KeyDLHQZZUEERVIGQ-UHFFFAOYNA-N
SMILESCCC(C)(O)CCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4310 to 1.4350 (20°C, 589 nm)
Appearance (Form)Liquid