4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Catalog number ACR104361000
Price (MYR)
503.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
503.00
Add to cart
Chemical Identifiers
CAS105-39-5
IUPAC Nameethyl 2-chloroacetate
Molecular FormulaC4H7ClO2
InChI KeyVEUUMBGHMNQHGO-UHFFFAOYSA-N
SMILESCCOC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Refractive Index1.4190-1.4230 @ 20?C
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless to pale yellow