4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR104330250
Price (MYR)
495.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
495.00
Add to cart
Specifications
Chemical Name or Material4-Aminophenyl ether
CAS101-80-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0