4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
CAS: 123-30-8 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number ACR104272500
Price (MYR)
467.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
467.00
Add to cart
Chemical Identifiers
CAS331717-63-6
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium; cyanosulfanide
Molecular FormulaC7H11N3S
InChI KeyVASPYXGQVWPGAB-UHFFFAOYSA-M
SMILES[S-]C#N.CCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR97.5% min