4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
CAS: 123-30-8 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number ACR104270050
Price (MYR)
444.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
444.00
Add to cart
Chemical Identifiers
CAS63139-21-9
IUPAC Name(4-ethylphenyl)boronic acid
Molecular FormulaC8H11BO2
InChI KeyRZCPLOMUUCFPQA-UHFFFAOYSA-N
SMILESCCC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)White to pale cream
Melting Point (clear melt)142.0-150.0?C
Proton NMRConforms to structure