4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
CAS: 123-30-8 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
250 g
Packaging:
Glass bottle
Catalog number ACR104270010
Price (MYR)
814.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
814.00
Add to cart
Specifications
Chemical Name or Material4-Aminophenol
CAS123-30-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0