2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
CAS: 95-55-6 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number ACR104261000
Price (MYR)
467.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
467.00
Add to cart
Chemical Identifiers
CAS109-43-3
IUPAC Name1,10-dibutyl decanedioate
Molecular FormulaC18H34O4
InChI KeyPYGXAGIECVVIOZ-UHFFFAOYSA-N
SMILESCCCCOC(=O)CCCCCCCCC(=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥90.0%
Refractive Index1.4375-1.4445 @ 20?C
Appearance (Color)Clear colorless