2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
CAS: 95-55-6 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR104260050
Price (MYR)
415.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
415.00
Add to cart
Chemical Identifiers
CAS75-50-3
IUPAC Nametrimethylamine
Molecular FormulaC3H9N
InChI KeyGETQZCLCWQTVFV-UHFFFAOYSA-N
SMILESCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration with HCl32 to 34 %