1-Aminoindan, 98%, Thermo Scientific Chemicals
1-Aminoindan, 98%, Thermo Scientific Chemicals
1-Aminoindan, 98%, Thermo Scientific Chemicals
1-Aminoindan, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Aminoindan, 98%, Thermo Scientific Chemicals
CAS: 34698-41-4 | C9H12N | 134.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR103850010
Price (MYR)
882.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
882.00
Add to cart
Specifications
Chemical Name or Material1-Aminoindan
Name Note99%
CAS34698-41-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning