2-Amino-5-chlorobenzoxazole, 97%, Thermo Scientific Chemicals
2-Amino-5-chlorobenzoxazole, 97%, Thermo Scientific Chemicals
2-Amino-5-chlorobenzoxazole, 97%, Thermo Scientific Chemicals
2-Amino-5-chlorobenzoxazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-chlorobenzoxazole, 97%, Thermo Scientific Chemicals
CAS: 61-80-3 | C7H5ClN2O | 168.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR103380050
Price (MYR)
724.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
724.00
Add to cart
Chemical Identifiers
CAS75277-39-3
IUPAC Namesodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular FormulaC8H17N2NaO4S
InChI KeyRDZTWEVXRGYCFV-UHFFFAOYSA-M
SMILES[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)≥98.5% (dry wt. basis)
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤3.0%
Appearance (Color)White