4-Aminobenzophenone, 98%, Thermo Scientific Chemicals
4-Aminobenzophenone, 98%, Thermo Scientific Chemicals
4-Aminobenzophenone, 98%, Thermo Scientific Chemicals
4-Aminobenzophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminobenzophenone, 98%, Thermo Scientific Chemicals
CAS: 1137-41-3 | C13H11NO | 197.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number ACR103170100
Price (MYR)
532.00
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
532.00
Add to cart
Chemical Identifiers
CAS14073-97-3
IUPAC Name(2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-one
Molecular FormulaC10H18O
InChI KeyNFLGAXVYCFJBMK-BDAKNGLRSA-N
SMILESCC(C)[C@@H]1CC[C@@H](C)CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=80 %
Specific optical rotation-2° to -10° (20°C, 589 nm) (c=4, ethanol)
Infrared spectrumConforms
Refractive index1.4490 to 1.4530 (20°C, 589 nm)