4-Aminoantipyrine, 98%, Thermo Scientific Chemicals
4-Aminoantipyrine, 98%, Thermo Scientific Chemicals
4-Aminoantipyrine, 98%, Thermo Scientific Chemicals
4-Aminoantipyrine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminoantipyrine, 98%, Thermo Scientific Chemicals
4-Aminoantipyrine, 98%, C11H13N3O, CAS Number-83-07-8, 4-aminoantipyrene, aminoantipyrin, ampyrone, 4-aminoantipyrine, 4-aminophenazone, metapirazone, solnapyrin-a, aminoazophene, solvapyrin-a, aminoantipyrine, 25g, (5% aq. soln.) clear, 24,273, 100.0, 97.5, CHEBI:59026, Brown-Yellow to Yellow
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
100 g
500 g
Packaging:
View all
Plastic Bottle
Catalog number ACR103150025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Aminoantipyrine
Name Note98%
CAS83-07-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning