4-Allylanisole, 98%, Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
CAS: 140-67-0 | C10H12O | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR102880250
Price (MYR)
499.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
499.00
Add to cart
Chemical Identifiers
CAS867-13-0
IUPAC Nameethyl 2-(diethoxyphosphoryl)acetate
Molecular FormulaC8H17O5P
InChI KeyGGUBFICZYGKNTD-UHFFFAOYSA-N
SMILESCCOC(=O)CP(=O)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Acid value=<2 mg KOH/g
Infrared spectrumConforms
Refractive index1.4300 to 1.4320 (20°C, 589 nm)
GC>=96.0 %