Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
CAS: 97-59-6 | C4H6N4O3 | 158.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ACR102845000
Price (MYR)
1,154.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,154.00
Add to cart
Chemical Identifiers
CAS12439-96-2
IUPAC Nameoxovanadiumbis(ylium) pentahydrate sulfate
Molecular FormulaH10O10SV
InChI KeyYGYBXHQARYQUAY-UHFFFAOYSA-L
SMILESO.O.O.O.O.O=[V++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<600 ppm
Titration with KMnO417 to 23 % (V)
Potassium (K)=<500 ppm
Appearance (Color)Blue
Degree of hydrationca. 5 moles water