Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
CAS: 97-59-6 | C4H6N4O3 | 158.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR102841000
Price (MYR)
499.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
499.00
Add to cart
Chemical Identifiers
CAS2892-51-5
IUPAC Namedihydroxycyclobut-3-ene-1,2-dione
Molecular FormulaC4H2O4
InChI KeyPWEBUXCTKOWPCW-UHFFFAOYSA-N
SMILESOC1=C(O)C(=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to beige
Titration with NaOH>=98.5 %