Adipic acid, 99%, Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
CAS: 124-04-9 | C6H10O4 | 146.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number ACR102815000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5020-41-7
IUPAC Name2-(3-methoxyphenyl)ethan-1-ol
Molecular FormulaC9H12O2
InChI KeyUPPGEJSCUZMCMW-UHFFFAOYSA-N
SMILESCOC1=CC=CC(CCO)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure