Adipic acid, 99%, Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
CAS: 124-04-9 | C6H10O4 | 146.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Plastic bottle
Catalog number ACR102810010
Price (MYR)
449.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
449.00
Add to cart
Chemical Identifiers
CAS98-74-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point75°C to 80°C
GC>=94 %