2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals
2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals
2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals
2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR102560250
Price (MYR)
517.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
517.00
Add to cart
Chemical Identifiers
CAS592-41-6
IUPAC Namehex-1-ene
Molecular FormulaC6H12
InChI KeyLIKMAJRDDDTEIG-UHFFFAOYSA-N
SMILESCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3867 to 1.3887 (20°C, 589 nm)