9-Acetylanthracene, 95%, Thermo Scientific Chemicals
9-Acetylanthracene, 95%, Thermo Scientific Chemicals
9-Acetylanthracene, 95%, Thermo Scientific Chemicals
9-Acetylanthracene, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9-Acetylanthracene, 95%, Thermo Scientific Chemicals
CAS: 784-04-3 | C16H12O | 220.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR102440050
Price (MYR)
510.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
510.00
Add to cart
Chemical Identifiers
CAS90-93-7
IUPAC Name4-[4-(diethylamino)benzoyl]-N,N-diethylaniline
Molecular FormulaC21H28N2O
InChI KeyVYHBFRJRBHMIQZ-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Melting Point (clear melt)91-97?C
Appearance (Color)Cream to yellow to green
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder