Check Order Status
Influenza B Goat anti-Human, Polyclonal, MilliporeSigma™
Actual product may vary
Thermo Fisher Scientific
Influenza B Goat anti-Human, Polyclonal, MilliporeSigma™
Goat Polyclonal Antibody
Have Questions?
Catalog number
AB-1058
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Detect Influenza B using this Anti-Influenza B Antibody validated for use in ELISA, immunofluoresence, western blot, immunohistochemistry.
For Research Use Only
Specifications
Antigen
Influenza B
Applications
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification
Polyclonal
Conjugate
Unconjugated
Formulation
IgG fraction. Liquid in PBS (0.01 M, pH 7.2) and 0.1% sodium azide. ≥95% pure.
Host Species
Goat
Immunogen
Yamagata strain of Influenza B
Quantity
1 Unit
Primary or Secondary
Primary
Target Species
Human
Form
Purified
Unit Size
Contents & Storage
2-8°C for 3 months or at −20°C in aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Store under subdued light.