Search Thermo Fisher Scientific
Catalog Number | Type | Teeth Count | Thickness (Metric) Teeth |
---|---|---|---|
A2-20D | Standard Comb | 20 | 1.5 mm |
A2-28C | Standard Comb | 28 | 1 mm |
A2-MT2D | Microwell Comb | 42 (2X) | 1.5 mm |
Eliminate comb assembly with an assortment of heavy-duty, one-piece Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems Combs, for use with Owl A2 and A5 Large Gel Electrophoresis Systems.