Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Actual product may vary
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Thermo Fisher Scientific
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Provide uncompromised sample protection for -50° to -86°C applications with Thermo ScientificTM FormaTM 900 Series -86°C Upright Ultra-Low Temperature Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrical RequirementsHoldsCapacity (English)No. of Doors
994230 V 50 HzBoxes (288)17.3 cu. ft.2
995230 V 50 HzBoxes (360)23 cu. ft.2
Catalog number 994
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Electrical Requirements:
230 V 50 Hz
Holds:
Boxes (288)
Capacity (English):
17.3 cu. ft.
No. of Doors:
2
Provide uncompromised sample protection for -50° to -86°C applications with Thermo ScientificTM FormaTM 900 Series -86°C Upright Ultra-Low Temperature Freezers. Available in single or double door models, these freezers are designed for daily sample protection and dependability.
Key Features
  • Centralized, eye level information center, including microprocessor control and monitoring system; all controls and displays are easy to reach and read
  • Improves temperature control and increases reserve BTUH capacity, resulting in a more stable temperature sample
  • Unique double door to store frequently used samples in the top chamber while long-term storage remains undisturbed in bottom
  • Single-hand operation: easy-to-use, padlock-compatible, ergonomic door handle with integrated key lock
  • Optional CO2 or LN2 back-up: Choice of CO2 or LN2 safety back-up systems for additional sample protection in the event of a power or mechanical failure
  • Optional chart recorder: Built-in chart recorder can be installed at eye level on Forma 900 -86°C models
Construction:
  • 5 in. (127mm) foamed-in-place, polyurethane insulation: Reduces power consumption, maintains temperature setpoint
  • Triple-point door gasket, provides longer holding time in the event of a door opening
  • Rugged, heavy gauge, cold-rolled steel cabinets resist chipping and rust
  • 1 in. (25mm) access ports: Allow for the use of inexpedient probes or instrumentation
  • Four inner doors reduce cold air loss and improve temperature recovery after door closings
  • Painted Galvaneel interior (stainless steel option available)
  • Water-cooled condenser option available
Warranty varies by country, please contact us for details
Specifications
DescriptionForma Series Ultra-Low Freezer, 288-Box Capacity, 230V, 50Hz
TypeUltra-Low Freezer, Upright
Capacity (English)17.3 cu. ft.
Capacity (Metric)288 L
Temperature Range-50°C to -86°C
Door StyleOuter (1), Inner (4)
No. of Doors2
Inner Doors4
Cabinet MaterialCold-rolled Steel
InsulationPolyurethane Foam
InteriorPainted Galvaneel - Standard, Stainless-steel - Optional
Shelves3
Capacity (English) Shelf150 lb.
Capacity (Metric) Shelf68 kg
Certifications/ComplianceCE, cULus, FDA Class I, UL
StyleUpright
DisplayPush Button LED
Condenser TypeFin and Tube, Forced-Air Cooled
Product LineForma
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Water CooledOptional
Dimensions (DxWxH) Exterior38.9 x 33.3 x 77.9 in. (98.9 x 84.6 x 197.9 cm)
Dimensions (D x W x H) Interior25.3 x 23 x 51.5 in. (64.3 x 58.4 x 130.8 cm)
Footprint (English)9.00 sq. ft.
Footprint (Metric).84 sq. meters
Monitoring OptionsAlarm, Recorder
Access SecurityKeyLock, Padlock Standard
Chart Recorder7-day Ink or Inkless (Optional)
Data OutputsRS-485, 4-20mA, dry contacts - standard, PT100 probe Optional
On-Board DataloggingNot Available
Operating ModesStandard Operation
Setpoint SecurityNot Available
Backup SystemsCO2 or LN2 Optional
Smart View CompatibilitySmart-Vue Compatible
Amperage12 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Wattage980 W
Energy Usage18kW/day
Line Voltage IndicatorBuck/Boost
Voltage CompensationBuck/Boost
Vial Capacity28,800 x 2mL
CompressorHermetic
RefrigerantR404A (1st stage), R508B (2nd stage)
Shipping Weight (English)763 lb.
Shipping Weight (Metric)346 kg
Weight (English)713 lb.
Weight (Metric)323 kg
HoldsBoxes (288)
Unit Size