KingFisher™ Plastics for 96 deep-well format
KingFisher™ Plastics for 96 deep-well format
Actual product may vary
Thermo Fisher Scientific
KingFisher™ Plastics for 96 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
9504045096 Deep Well plate, v-bottomKF Duo Prime, Flex and Presto
97003530Combi PackKingFisher Duo Prime only
Catalog number 95040450
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
96 Deep Well plate, v-bottom
For Use With (Equipment):
KF Duo Prime, Flex and Presto
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Product Type96 Deep Well plate, v-bottom
Format96 Deep-well Plate
Capacity96 Samples
DescriptionKingFisher 96 deep-well plate, v-bottom, polypropylene (for Duo Prime, Flex and Presto)
For Use With (Equipment)KF Duo Prime, Flex and Presto
Quantity50/Pk.
Unit Size