Finntip™ Flex™ Pipette Tips
Finntip™ Flex™ Pipette Tips
Actual product may vary
Thermo Fisher Scientific
Finntip™ Flex™ Pipette Tips
The Thermo Scientific™ Finntip™ Flex™ Pipette Tips reduce the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI). Ensure a better seal with the pipetter module, for higher accuracy and precision.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Color CodeFit for Purpose
94060520300 μLOrangeCertified Quality
9406012010 μLPinkCertified Quality
9406010010 μLPinkCertified Quality
94060310200 μLYellowCertified Quality
9406010310 μLPinkSterile & PCR Ready
94060313200 μLYellowSterile & PCR Ready
94060320200 μLYellowCertified Quality
94060510300 μLOrangeCertified Quality
94060513300 μLOrangeSterile & PCR Ready
940607101000 μLBlueCertified Quality
940607131000 μLBlueSterile & PCR Ready
940607201000 μLBlueCertified Quality
Catalog number 94060520
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
300 μL
Color Code:
Orange
Fit for Purpose:
Certified Quality
Finntip™ Flex™ Pipette Tips are designed to fit Finnpipette™ F1, F2 and Novus pipettes. They enable you to pipette accurately and precisely, with low liquid retention. The compatibility between the tip and the pipette is indicated with color codes. These Flex™ tips have an ergonomic design that lower the risk of RSI and use less plastic.
Key Features:
  • Excellent for single and multichannel pipetting
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized, 10 x 96 rack
Softer Finntip Flex Tip
  • The softer tip collar reduces the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI)
  • It also ensures a better seal with the pipetter module, ensuring higher accuracy and precision
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • Available in six volume ranges: 0.2-10 μL, 1-200 μL, 5-300 μL, 100-1000 μL, 50-1200 μL and 10mL Extended (bottle tip)
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip Flex pipette tip rack insert
  • Color-coding visible in rack and reload packaging options
Note:
Availability may vary by country.
Specifications
DescriptionFinntip Flex 300, Non-Filtered, Non-sterile, Bulk
Volume (Metric)300 μL
FilteredNon-filtered
SterilityNon-sterile
ColorOrange
ModelFinntip Flex 300
Length (English)2.42 in.
Length (Metric)6.15cm
FormatBulk
Packaging1000/Bag
Color CodeOrange
For Use With (Equipment)Thermo Scientific Finnpipette
Product LineFinnTip Flex
Fit for PurposeCertified Quality
Unit Size960