Search Thermo Fisher Scientific
| Catalog Number | Description | For Use With (Equipment) | Holds |
|---|---|---|---|
| 920017 | |||
| 830032 | |||
| 920018 | 2 in. Box Racks - Adjustable side access rack | 17.3 and 23 cu. ft. capacity upright freezers | Boxes (2 in.) |
| 920022 | |||
| 920094 | |||
| 920096 | 2 in. Box Racks - Sliding drawer rack | 17.3 and 23 cu. ft. capacity upright freezers | pk |
| 920097 |
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.