S1 Pipet Filler Accessories
S1 Pipet Filler Accessories
Actual product may vary
Thermo Fisher Scientific
S1 Pipet Filler Accessories
Incorporate Thermo Scientific™ S1 Pipet Filler Accessories with S1 Pipet Filler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
9067Table Mount Stand
4580560Pipette Filter, Non-sterile
FIS14-386-77Pipette Filter, Non-sterile
9057Pipette Filter, Sterile
FKS10193923Pipette Filter, Sterile
9065-11Silicone Pipet Gripper
9069Wing Stand
Catalog number 9067
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Table Mount Stand
Complete your S1 Pipet Filler with these accessories and prolong its use.
Features:
  • Wall-mounted holder and table stand offer compact storage between applications. Recharge the pipet filler in both stands by plugging in the charger. Store pipet filler when working in laminar flow cabinets to save space.
  • Wing stand allows user to set the pipet filler down without removing or emptying the serological pipet.
  • Easy to change filter prevents aerosol or liquid cross contamination of the pipet together with autoclavable silicone adapter.
  • 1mL Pipet Support ensures that 1 mL pipets will hold as securely as larger sizes.
Note:
Wing stand not available for use or sale within the United States or for importation into the United States.
Specifications
For Use WithS1 Pipette Filler
TypeTable Mount Stand
For Use With (Equipment)Single Pipet Filler
Unit SizeEACH