Racks and Accessories for CO2 Resistant Shaker
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Actual product may vary
Thermo Fisher Scientific
Racks and Accessories for CO2 Resistant Shaker
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
88881123Rubber mat platform
88881119Fixed Tube Rack 40 x 16 mm dia.
88881122Universal aluminum platform
8888112496-Well microplate holder
88881126Sticky mat 140 x 140 mm
88881132UK Plug
Catalog number 88881123
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Includes:
Rubber mat platform
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
  • Compatible with Thermo Scientific™ CO2 Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Product TypeRubber Platform
For Use With (Equipment)CO2 Resistant Shaker
IncludesRubber mat platform
Unit Size