Clamps for CO2 Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Actual product may vary
Clamps for CO<sub>2</sub> Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Thermo Fisher Scientific
Clamps for CO2 Resistant Shaker
Achieve maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
Have Questions?
Catalog NumberClampsProductTypeSpecs
88881112
Catalog number 88881112
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
  • Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
For Use With (Equipment)CO2 Resistant Shaker
No. of Positions4
Unit Size