Clamps for CO2 Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Actual product may vary
Clamps for CO<sub>2</sub> Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Thermo Fisher Scientific
Clamps for CO2 Resistant Shaker
Achieve maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClampsProduct Type
88881110500 mLFlask Clamp
88881107150 mLFlask Clamp
88881109250 mLFlask Clamp
888811111000 mL Flask Clamp
888811122000 mLFlask Clamp
88881133FlaskStarter Kit
Catalog number 88881110
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Clamps:
500 mL
Product Type:
Flask Clamp
Maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
  • Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Product TypeFlask Clamp
Clamps500 mL
For Use With (Equipment)CO2 Resistant Shaker
No. of Positions9
Unit Size