Accessories for RT2 Hotplate, RT2 Basic and Advanced Hotplate Stirrers
Accessories for RT2 Hotplate, RT2 Basic and Advanced Hotplate Stirrers
Actual product may vary
Thermo Fisher Scientific
Accessories for RT2 Hotplate, RT2 Basic and Advanced Hotplate Stirrers
Use these accessories to optimize the performance of your Thermo Scientific™ RT2 Hotplate, RT2 Basic and Advanced Hotplate Stirrers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
88880148C-5, Clamp Holder (PP body, 12 mm), Available for RT2 Advanced Hotplate Stirrer only
88880141Heating Bath
88880143Support Rod (Ø12mm, 400mm, M10)
88880142Transparent Shield (PC)
88880147Temperature Probe (PT 100, SN-8-4 connector sensor), Available for RT2 Advanced Hotplate Stirrer only
888801493 Prong Clamp (60mm grip), Avaialble for RT2 Advanced Hotplate Stirrer only
88880150Temperature Probe (PT100, A Class, 400°C), Avaialble for RT2 Advanced Hotplate Stirrer only
Catalog number 88880148
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
C-5, Clamp Holder (PP body, 12 mm), Available for RT2 Advanced Hotplate Stirrer only
Use these accessories to optimize the performance of your Thermo Scientific™ RT2 Hotplate, RT2 Basic and Advanced Hotplate Stirrers.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionC-5, Clamp Holder (PP body, 12 mm), Available for RT2 Advanced Hotplate Stirrer only
For Use With (Equipment)RT2 Series Hotplate/Stirrers
TypeClamp
Unit Size