Optimize the performance of our digital heating cooling drybath and digital heating shaking bath by using these accessories.
Have Questions?
Change view
Catalog Number
No. of Places
Tube Size
Type
88880131
15
15mL
Block
88880130
30
1.5mL
Block
88880133
6
50mL large
Block
3 Options
Catalog number 88880131
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
No. of Places:
15
Tube Size:
15mL
Type:
Block
Use Thermo Scientific™ Accessories for Digital Heating Cooling Bath to optimize flexibility and performance.
Warranty: 2 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (Metric)15 mL
LightingLED
No. of Places15
TypeBlock
Tube Size15mL
Dimensions (L x W x H)3 x 3.9 x 2 in. (76.5 x 98 x 51 mm)