Use Thermo Scientific Accessories for Digital Shaking Baths to optimize flexibility and performance.
Have Questions?
Change view
Catalog Number
No. of Places
Tube Size
88880126
48
0.5mL
88880125
96
0.2mL
88880127
24
1.5mL
88880128
6
50mL
4 Options
Catalog number 88880126
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
No. of Places:
48
Tube Size:
0.5mL
Use Thermo Scientific Accessories for Digital Heating Shaking Baths to optimize flexibility and performance.
Warranty: 2 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.