Accessories for Digital Heating Shaking Baths
Accessories for Digital Heating Shaking Baths
Actual product may vary
Thermo Fisher Scientific
Accessories for Digital Heating Shaking Baths
Use Thermo Scientific Accessories for Digital Shaking Baths to optimize flexibility and performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of PlacesTube Size
88880125960.2mL
88880126480.5mL
88880127241.5mL
88880128650mL
Catalog number 88880125
View Price:Sign in to see your account pricing. Need an account? Register with us today.
No. of Places:
96
Tube Size:
0.2mL
Use Thermo Scientific Accessories for Digital Heating Shaking Baths to optimize flexibility and performance.
  • Warranty: 2 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (Metric)0.2 mL
For Use With (Equipment)Heating/Cooling Bath
LightingLED
No. of Places96
TypeAccessory, Block Set with Cover
Tube Size0.2mL
Unit Size