Block, 24 x 0.2 mL
Product Image
Thermo Fisher Scientific
Block, 24 x 0.2 mL
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Have Questions?
Catalog number 88871101
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 years
Specifications
Description24 x 0.2mL block
Capacity (Metric)0.2 mL
For Use With (Equipment)Thermo Scientific™ Compact Dry Bath/Block Heater
LightingLED
No. of Places24
TypeBlock
Dimensions (L x W x H)36 x 54 x 15 mm (1.4 x 2.1 x 0.6 in.)
Unit Size