Block, 15 mL Conical Tube
Block, 15 mL Conical Tube
Actual product may vary
Thermo Fisher Scientific
Block, 15 mL Conical Tube
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog number 88870118
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
  • Warranty: 2 years
Specifications
Description15mL conical tube block
Capacity (Metric)15 mL
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds15 x 15mL conical
LightingLED
No. of Places15
TypeBlock
Dimensions (L x W x H)76 x 124 x 113 mm (3 x 4.9 x 4.55 in.)
Unit Size