Block, 22 mm, 13 mm and 6 mm dia.
Block, 22 mm, 13 mm and 6 mm dia.
Actual product may vary
Thermo Fisher Scientific
Block, 22 mm, 13 mm and 6 mm dia.
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog number 88870114
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
  • Warranty: 2 years
Specifications
DescriptionMixed block, 22mm, 13mm and 6mm dia.
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds3 x 25mm, 12 x 12∽13mm and 6 x 6mm
LightingLED
TypeBlock
Dimensions (L x W x H)76 x 124 x 39 mm (3 x 4.9 x 1.5 in.)
Unit SizeEA