GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
Actual product may vary
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
Thermo Fisher Scientific
GENESYS™ 30 Visible Spectrophotometer Accessories
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
840-278500Replacement Compartment Lid
840-278000Thermal Snap-on Printer
840-259800Self-Stick Printer Paper
840-278300Standard Printer Paper
840-278700Tungsten Halogen Lamp
Catalog number 840-278500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Replacement Compartment Lid
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible In addition to valuable built-in features such as the high resolution color display, large sample compartment, and data export options, a complete selection of accessories are available for the the GENESYS 30 spectrophotometer to help meet the needs of students, educators and lab technicians for accurate quantitative analysis.
  • Holds one rectangular longpath cell 20 - 100mm pathlength 
  • Comes assembled in a removable sample compartment liner
  • Allows for flexibility between samples
Specifications
TypeReplacement Compartment Lid
For Use With (Application)Replace your spectrophotometer lid and hinge assembly yourself if it is broken through abuse.
For Use With (Equipment)GENESYS 30 Visible Spectrophotometer
Includes1 lid and hinge assembly
Storage RequirementsAmbient
Warranty2 Years
Unit Size