Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Have Questions?
Change view
Catalog Number
Type
840-259800
Self-Stick Printer Paper
840-278000
840-278300
840-278700
840-278500
5 Options
Catalog number 840-259800
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Type:
Self-Stick Printer Paper
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible In addition to valuable built-in features such as the high resolution color display, large sample compartment, and data export options, a complete selection of accessories are available for the the GENESYS 30 spectrophotometer to help meet the needs of students, educators and lab technicians for accurate quantitative analysis.
Holds one rectangular longpath cell 20 - 100mm pathlength
Comes assembled in a removable sample compartment liner
Allows for flexibility between samples
Specifications
TypeSelf-Stick Printer Paper
For Use With (Application)Print your samples and adhere them to your lab booklet without the need to staples or tape.
For Use With (Equipment)Thermal printers for: GENESYS 30 and 10S, BioMate 3S, Evolution 60S, SPECTRONIC 200/200E