Search Thermo Fisher Scientific
Catalog Number | Capacity | For Use With (Equipment) |
---|---|---|
75007304 | 4 x 750 mL | 750 mL Polypropylene Wide-Mouth Bio-Bottle and any TX-750 Round adapter |
75003829 | 150 to 300 mL Small Blood Bags | Small bag/cell culture bags 4 x 2 bags (< 350mL) |
75007301 | 4 x 1000 mL | 1000mL Bio-Bottle |
75004252 | 48 x 50 mL | 50mL Thermo Scientific Nalgene Oak Ridge Tube |
75004253 | 4 x 500 mL | 500mL Nalgene Bottle |
75005392 | 4 x 250 mL | 250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle |
75007302 | 4 x 500 mL | 500mL Corning Bottle |
75007303 | 4 x 6 mL Standard or 4 x 2 mL Deep-Well | Microplate carriers |
75007305 | 8 x 250 mL | 250mL Thermo Scientific Nalgene Bottle |
75007306 | 96 x 15 mL | 15mL Conical Tube |