Check Order Status
Adapters for HIGHConic III Rotor
Actual product may vary
Thermo Fisher Scientific
Adapters for HIGHConic III Rotor
Assorted adapters to be used with the HIGHConic III rotor
Have Questions?
Change view
Catalog Number
Capacity
Max. Tube Diameter
Max. Tube Length
75005807
75005802
75005805
6.5 mL
13.5 mm
114 mm
75005808
8 x 15 mL
12 mm
17 mm
75005770
5 Options
Catalog number
75005807
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Accommodate conical tubes in the 50mL buckets for use with the Thermo Scientific HIGHConic III fixed angle rotor.
Adapters to run various tubes in HIGHConic III rotor
Specifications
No. of Places
2
Unit Size