Adapters for HIGHConic III Rotor
Adapters for HIGHConic III Rotor
Actual product may vary
Thermo Fisher Scientific
Adapters for HIGHConic III Rotor
Assorted adapters to be used with the HIGHConic III rotor
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityMax. Tube DiameterMax. Tube Length
750057705 mL17 mm100 mm
7500580238 mL25.5 mm110 mm
750058056.5 mL13.5 mm114 mm
750058071.5/2.0 mL11 mm40 mm
750058088 x 15 mL12 mm17 mm
Catalog number 75005770
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
5 mL
Max. Tube Diameter:
17 mm
Max. Tube Length:
100 mm
Accommodate conical tubes in the 50mL buckets for use with the Thermo Scientific HIGHConic III fixed angle rotor.
  • Adapters to run various tubes in HIGHConic III rotor
Specifications
TypeRotor Adapter
Tube TypeRound Bottom
MaterialPolypropylene
Capacity5 mL
Max. Tube Diameter17 mm
Max. Tube Length100 mm
For Use With (Equipment)HIGHConic III rotor (75005709)
DescriptionAdapters for 5mL round bottom tubes used in HIGHConic III rotor
No. of Places1
Tube Size5 mL
No. per Pack2
Unit Size