Accessories for Thermo Scientific™ HAEMAFlex™ Rotors
Actual product may vary
Thermo Fisher Scientific
Accessories for Thermo Scientific™ HAEMAFlex™ Rotors
Complete your rotor system with accessories for Thermo Scientific™ HAEMAFlex™ rotors.
Have Questions?
Change view
Catalog Number
For Use With (Equipment)
Type
75003843
HAEMAFlex Swinging Bucket Rotor
Blood Bag Spacer
75003866
HAEMAFlex Swinging Bucket Rotor
Dummy Weight
2 Options
Catalog number 75003843
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
HAEMAFlex Swinging Bucket Rotor
Type:
Blood Bag Spacer
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Accessories designed to complete your HAEMAFlex rotor system.
Specifications
TypeBlood Bag Spacer
For Use With (Equipment)HAEMAFlex Swinging Bucket Rotor