Check Order Status
Centrifugation Polypropylene Bio-Bottles
Actual product may vary
Thermo Fisher Scientific
Centrifugation Polypropylene Bio-Bottles
Use these polypropylene screw cap closure bio-bottles for compatibility with round-bucket centrifuge systems.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
75003800
180 mL
75007585
400 mL
FKS10482525
400 mL
3 Options
Catalog number
75003800
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
180 mL
Use these polypropylene screw cap closure bio-bottles for centrifugation compatibility with round buckets as in the Thermo Scientific™ TX-400 Rotor.
With screw cap
The 180mL Bio-Bottles
(75-003-800)
are compatible with Round Buckets for Thermo Scientific TX-200 Rotor
(75-003-569)
The 400mL Bio-Bottles
(75-007-585)
are compatible with Round Buckets for Thermo Scientific TX-400 Rotor
(75-003-655)
Specifications
Capacity
12 x 180 mL
Capacity (Metric)
180 mL
Color
Clear
Material
Polypropylene
Shape
Round
Product Type
Centrifuge Bottle
For Use With (Equipment)
Round Bucket for Thermo Scientific TX-200 Rotor
Closure Material
Polypropylene
Closure Type
Screw Cap
Unit Size