Fiberlite™ F13-14 x 50cy Fixed Angle Rotor PROMO
Fiberlite™ F13-14 x 50cy Fixed Angle Rotor PROMO
Actual product may vary
Fiberlite™ F13-14 x 50cy Fixed Angle Rotor PROMO
Fiberlite™ F13-14 x 50cy Fixed Angle Rotor PROMO
Thermo Fisher Scientific
Fiberlite™ F13-14 x 50cy Fixed Angle Rotor PROMO
Fiberlite™ F13-14 x 50cy Fixed Angle Rotor provides 14-place capacity and high maximum RCF.
Have Questions?
Catalog NumberdimCapacity
75003661
Catalog number 75003661
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Capacity: 14 × 50 mL
  • Maximum Speed/RCF: 10,000 rpm/17,105 x g†
Fiberlite Rotors
These lightweight, carbon fiber rotors maximize performance with versatility, speed and a robust, corrosion-free design.
  • Enhanced productivity: Reduce run-times with higher G-forces with our lightweight carbon fiber rotors
  • Unequalled durability: Corrosion-and fatigue-resistant design ensures structural integrity
  • Secure investment: Backed by a 15-year warranty
  • Compatible with Thermo Scientific Nalgene™ bottles and tubes for complete centrifuge solutions
Ordering Information:
Tube adapters are sold separately. For O-ring refresher kit, order 021-149-027 or contact your Sales Representative for more information.
Compatible with:
Thermo Scientific Multifuge™ X3/X3R/X3F/X3FR, Sorvall™ Legend™ XT/XF/XTR Centrifuge Series.
Recommended for:
Pelleting prokaryotic and eukaryotic cells from cell cultures.
Note:
†Performance may vary by centrifuge model
Specifications
Radius (Metric) Min.8.3 cm
Unit Size