CLINIConic Fixed-Angle Rotor for Compact and 1.6 L General Purpose X Pro and ST Plus Centrifuges
CLINIConic Fixed-Angle Rotor for Compact and 1.6 L General Purpose X Pro and ST Plus Centrifuges
Actual product may vary
CLINIConic Fixed-Angle Rotor for Compact and 1.6 L General Purpose X Pro and ST Plus Centrifuges
CLINIConic Fixed-Angle Rotor for Compact and 1.6 L General Purpose X Pro and ST Plus Centrifuges
Thermo Fisher Scientific
CLINIConic Fixed-Angle Rotor for Compact and 1.6 L General Purpose X Pro and ST Plus Centrifuges
Support a range of capacities and applications by using the Thermo Scientific™ CLINIConic™ Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
7500362330 x 15 mL
Catalog number 75003623
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
30 x 15 mL
Optimize separations with fast acceleration of 30 blood tubes or 15mL conicals in a fixed angle configuration with the Thermo Scientific™ CLINIConic™ Fixed Angle Rotor.
Spin 30 blood tubes or 15mL conicals in a fixed angle configuration with fast acceleration to optimize applications.
Ordering Information:
Tube adapters are sold separately; order rotor body (75-003-623) and components to complete assembly. Contact your sales representative for information.
Specifications
DescriptionCLINIConic Fixed-Angle Rotor
TypeFixed Angle Rotor
Capacity30 x 15 mL
Max. RCF4,997 x g
Max. Speed5,650 rpm
Radius (Metric)140 mm
Max. Tube Length131 mm
Max. Tube Diameter16.5 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Sorvall ST 8, Multifuge X1, Megafuge 16, Megafuge 8, SL 16 and SL 8 Centrifuges
Angle37°
Unit Size