24 x 1.5/2.0mL Fixed Angle Rotor
24 x 1.5/2.0mL Fixed Angle Rotor
Actual product may vary
Thermo Fisher Scientific
24 x 1.5/2.0mL Fixed Angle Rotor
Biocontainment-approved, autoclavable aluminum rotor with cover
Have Questions?
Catalog NumberCapacity
7500333224 x 1.5/2 mL
Catalog number 75003332
Price (MYR)
5,331.14
Add to cart
Capacity:
24 x 1.5/2 mL
Price (MYR)
5,331.14
Add to cart
  • Capacity: Four microplates or two deepwell plates
  • Max. Speed/RCF: 18,000rpm/21,890 x g
  • Max. Tube Size: 24 × 1.5 to 2.0mL
Specifications
TypeFixed Angle Rotor
Capacity24 x 1.5/2 mL
Max. RCF28,110 x g
Max. Speed15,000 rpm
Radius (Metric)0.87 cm
For Use With (Equipment)Thermo Scientific™ Sorvall™ Stratus Centrifuge
Angle45°
Certifications/ComplianceBiocontainment approved