H-Flex 1 Swinging Bucket Rotor and Accessories
H-Flex 1 Swinging Bucket Rotor and Accessories
Actual product may vary
Thermo Fisher Scientific
H-Flex 1 Swinging Bucket Rotor and Accessories
Spin more with less! High capacity and flexibility—all in a small footprint.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
7500330533 x 10/12 mL Adapter
7500330418 x 15 mL Adapter
7500330333 x 10/12 mL Adapter
75003300Cross
75003301Buckets
75003302Biocontainment Lids
75003307Microplate Adapter
Catalog number 75003305
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
33 x 10/12 mL Adapter
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
  • Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
  • Spins blood tubes, conical tubes and microplates without changing rotor or buckets
  • Get results faster without the need to change hardware
  • Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Specifications
Description33 x 10/12 mL Adapter
MaterialMolded Polyamide Nylone
TypeSwinging Bucket Rotor
Capacity33 x 10/12 mL Blood Tubes Each Adapter
Max. Speed4297
For Use With (Equipment)75003300 H-Flex Rotor Cross and 75003301 Buckets
BiocontainmentYes
AutoclavableAutoclavable
QuantitySet of 2
Unit Size